Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033573.1 | NP_001028745.1 | 3 | 845 | ||
NM_025883.3 | NP_080159.3 | 3 | 729 | |||
GenBank mRNA | AK010296.1 | - | 3 | 724 | ||
AK011373.1 | - | 3 | 696 | |||
AK035172.1 | - | 808 | ||||
AK038177.1 | - | 708 | ||||
AK050388.1 | - | 3 | 844 | |||
AK077693.1 | - | 3 | 683 | |||
AK077838.1 | - | 694 | ||||
AK139707.1 | - | 3 | 686 | |||
AK144128.1 | - | 3 | 701 | |||
AK154741.1 | - | 3 | 707 | |||
AK157494.1 | - | 3 | 711 | |||
AK159304.1 | - | 3 | 730 | |||
AK168497.1 | - | 3 | 724 | |||
BC033317.1 | - | 3 | 676 | 4459242 |