Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025892.2 | NP_080168.1 | 4 | 763 | ||
GenBank mRNA | AK005324.1 | - | 4 | 763 | ||
AK007713.1 | - | 4 | 747 | |||
BC023715.1 | - | 3 | 443 | 5346460 |