Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035509.1 | NP_001030586.1 | 4 | 884 | ||
NM_001035510.1 | NP_001030587.1 | 2 | 1355 | |||
NM_025893.2 | NP_080169.2 | 4 | 832 | |||
GenBank mRNA | AK005317.1 | - | 4 | 810 | ||
AK075607.1 | - | 1 | 436 | |||
AK134856.1 | - | 1213 | ||||
AY650116.1 | - | 1 | 314 | |||
BC017627.1 | - | 4 | 745 | 4505033 |