Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025894.2 | NP_080170.1 | 2 | 169 | ||
GenBank mRNA | AK005114.1 | - | 2 | 204 | ||
AK007619.1 | - | 2 | 214 | |||
AK007680.1 | - | 2 | 143 | |||
AK008646.1 | - | 2 | 170 | |||
AK009969.1 | - | 2 | 172 | |||
AK011806.1 | - | 2 | 193 | |||
AK017843.1 | - | 2 | 186 | |||
AK150910.1 | - | 2 | 193 | |||
AK151242.1 | - | 2 | 170 | |||
AK152041.1 | - | 2 | 193 | |||
AK152240.1 | - | 2 | 201 | |||
AK160283.1 | - | 2 | 191 | |||
AK162779.1 | - | 170 | ||||
AK167236.1 | - | 2 | 177 | |||
BC004694.1 | - | 2 | 187 | 3498952 |