Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025903.2 | NP_080179.1 | 3 | 401 | ||
GenBank mRNA | AK007687.1 | - | 3 | 402 | ||
AK149496.1 | - | 3 | 382 | |||
AK156625.1 | - | 3 | 395 | |||
AK165967.1 | - | 3 | 394 | |||
AK166130.1 | - | 3 | 469 | |||
AK166258.1 | - | 3 | 359 | |||
AY298860.1 | - | 3 | 230 | |||
BC023486.1 | - | 3 | 349 | |||
BC037434.1 | - | 3 | 357 | 2651172 |