Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025910.3 | NP_080186.3 | 2 | 484 | ||
GenBank mRNA | AB177385.1 | - | 1 | 285 | ||
AK007810.1 | - | 2 | 454 | |||
AK013451.1 | - | 2 | 444 | |||
AK014426.1 | - | 2 | 438 | |||
AK031671.1 | - | 2 | 418 | |||
AK032595.1 | - | 2 | 455 | |||
BC023462.1 | - | 2 | 739 | 5041558 | ||
BC025109.1 | - | 2 | 406 | 4236994 | ||
BC058242.1 | - | 2 | 393 | 6308890 |