Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000016.5 | NP_000007.1 | 7, 8 | 1035 | ||
NM_001127328.2 | NP_001120800.1 | 7, 8 | 1047 | |||
NM_001286042.1 | NP_001272971.1 | 6, 7 | 947 | |||
NM_001286043.1 | NP_001272972.1 | 8, 9 | 1134 | |||
NM_001286044.1 | NP_001272973.1 | 4, 5 | 765 | |||
GenBank mRNA | AF251043.1 | - | 7, 8 | 614 | ||
AK296045.1 | - | 4, 5 | 345 | |||
AK301005.1 | - | 4, 5 | 503 | |||
AK301039.1 | - | 7, 8 | 691 | |||
AK301717.1 | - | 6, 7 | 539 | |||
AK311022.1 | - | 6 | 516 | |||
AK312629.1 | - | 7, 8 | 625 | |||
AK315946.1 | - | 6, 7 | 588 | |||
BC005377.1 | - | 7, 8 | 629 | 3934299 | ||
BX647734.1 | - | 8, 9 | 773 | |||
KU177876.1 | - | 7, 8 | 594 | |||
KU177877.1 | - | 6, 7 | 506 | |||
KU177878.1 | - | 3, 4 | 237 | |||
M16827.1 | - | 7, 8 | 612 |