Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam210b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025912.4 | NP_080188.3 | 3 | 746 | ||
GenBank mRNA | AK008190.1 | - | 3 | 687 | ||
AK076087.1 | - | 3 | 689 | |||
AK084415.1 | - | 3 | 699 | |||
AK147935.1 | - | 3 | 705 | |||
AK148213.1 | - | 3 | 697 | |||
AK148334.1 | - | 3 | 687 | |||
AK148435.1 | - | 3 | 687 | |||
AK149497.1 | - | 3 | 684 | |||
AK160529.1 | - | 3 | 747 | |||
BC016210.1 | - | 3 | 719 | 4924329 | ||
BC057642.1 | - | 3 | 706 | 2655111 |