Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025920.3 | NP_080196.3 | 3 | 1601 | ||
GenBank mRNA | AK086483.1 | - | 3 | 1415 | ||
AK140888.1 | - | 1544 | ||||
AK167814.1 | - | 3 | 1361 | |||
AK213476.1 | - | 2 | 260 | |||
AK216112.1 | - | 2 | 322 | |||
BC013538.1 | - | 2 | 438 | |||
BC057963.1 | - | 2 | 899 | |||
BC063758.1 | - | 3 | 1332 | 6334411 | ||
BC066042.1 | - | 3 | 1536 | 6413779 |