Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025928.3 | NP_080204.1 | 2 | 221 | ||
GenBank mRNA | AF348510.1 | - | 2 | 231 | ||
AK011174.1 | - | 2 | 223 | |||
AK012113.1 | - | 2 | 222 | |||
AK013787.1 | - | 2 | 224 | |||
AK078308.1 | - | 2 | 240 | |||
AK150603.1 | - | 2 | 238 | |||
AK168200.1 | - | 2 | 222 | |||
BC034333.1 | - | 2 | 199 | |||
BC037139.1 | - | 2 | 230 | 4952550 | ||
BC049223.1 | - | 2 | 220 | 3257320 |