Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025936.3 | NP_080212.2 | 3 | 244 | ||
GenBank mRNA | AK011383.1 | - | 3 | 222 | ||
AK076160.1 | - | 3 | 219 | |||
AK149856.1 | - | 3 | 219 | |||
AK151966.1 | - | 2 | 42 | |||
AK152108.1 | - | 3 | 219 | |||
AK160216.1 | - | 3 | 219 | |||
AK166097.1 | - | 3 | 219 | |||
AK168194.1 | - | 3 | 219 | |||
BC020132.1 | - | 3 | 202 | 3989952 | ||
CF748175.1 | - | 3 | 245 |