Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190409.1 | NP_001177338.1 | 2 | 264 | ||
NM_025952.4 | NP_080228.4 | 2 | 264 | |||
GenBank mRNA | AK010320.1 | - | 2 | 200 | ||
AK012185.1 | - | 2 | 141 | |||
AK013243.1 | - | 2 | 145 | |||
AK018623.1 | - | 2 | 167 | |||
AK136622.1 | - | 2 | 146 | |||
AK144696.1 | - | 2 | 143 | |||
BC003881.1 | - | 2 | 151 | 3496572 | ||
DQ000005.1 | - | 2 | 264 |