Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025974.2 | NP_080250.1 | 4, 5 | 329 | ||
GenBank mRNA | AK002946.1 | - | 4, 5 | 330 | ||
AK007807.1 | - | 4, 5 | 330 | |||
AK010600.1 | - | 4, 5 | 332 | |||
AK012277.1 | - | 4, 5 | 330 | |||
AK013912.1 | - | 4, 5 | 330 | |||
AK141887.1 | - | 3, 4 | 887 | |||
AK167146.1 | - | 4, 5 | 330 | |||
BC013249.1 | - | 1, 2 | 88 | |||
BC013251.1 | - | 1, 2 | 88 | |||
BC037202.1 | - | 3, 4 | 330 | |||
BC042937.1 | - | 3, 4 | 308 |
Gene Symbol: Rpl14-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC008177.1 | - | 1 | 178 | ||
BC092249.1 | - | 1 | 314 | 5034997 |