Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025976.5 | NP_080252.1 | 2, 3 | 566 | ||
GenBank mRNA | AK013874.1 | - | 2, 3 | 431 | ||
AK041839.1 | - | 2, 3 | 475 | |||
AK178133.1 | - | 1, 2 | 278 | |||
BC027221.1 | - | 2, 3 | 407 | 4196466 |