Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrarp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025980.2 | NP_080256.2 | 1 | 977 | ||
GenBank mRNA | AK012426.1 | - | 1 | 713 | ||
AY046077.1 | - | 1 | 977 | |||
BC020293.1 | - | 1 | 974 | |||
BC069891.1 | - | 1 | 730 | 5391567 |