Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025878.1 | NP_080154.1 | 7 | 959 | ||
GenBank mRNA | AK002554.1 | - | 4 | 704 | ||
AK010250.1 | - | 7 | 966 | |||
AK076033.1 | - | 4 | 704 | |||
AK076044.1 | - | 4 | 704 | |||
AK088540.1 | - | 7 | 955 | |||
AK166597.1 | - | 7 | 980 | |||
AK167067.1 | - | 7 | 960 | |||
AK168140.1 | - | 7 | 974 | |||
BC021752.1 | - | 7 | 959 | 5136720 |