Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168475.1 | NP_001161947.1 | 6 | 1097 | ||
NM_001168476.1 | NP_001161948.1 | 5 | 897 | |||
NM_025905.3 | NP_080181.2 | 6 | 1097 | |||
GenBank mRNA | AK005431.1 | - | 5 | 858 | ||
AK076802.1 | - | 5 | 691 | |||
AK086049.1 | - | 1164 | ||||
AK141754.1 | - | 6 | 1083 | |||
BC038179.1 | - | 6 | 1068 | 4485073 |