Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam210b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025912.4 | NP_080188.3 | 3 | 3033 | ||
GenBank mRNA | AK147935.1 | - | 3 | 2992 | ||
AK148334.1 | - | 3 | 2971 | |||
AK149497.1 | - | 3 | 2971 | |||
BC016210.1 | - | 3 | 2987 | 4924329 |