Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025922.2 | NP_080198.2 | 8 | 1075 | ||
GenBank mRNA | AB100501.1 | - | 8 | 1087 | ||
AK008279.1 | - | 8 | 1115 | |||
AK009248.1 | - | 8 | 1086 | |||
AK153904.1 | - | 8 | 1094 | |||
BC026508.1 | - | 8 | 1075 | 4512398 | ||
BC094466.1 | - | 8 | 1063 | 4936811 |