Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025926.4 | NP_080202.1 | 3 | 1135 | ||
NM_027287.4 | NP_081563.2 | 4 | 1157 | |||
GenBank mRNA | AF157598.1 | - | 3 | 859 | ||
AK006478.1 | - | 4 | 1157 | |||
AK008537.1 | - | 3 | 1048 | |||
AK028049.1 | - | 3 | 1044 | |||
AK136240.1 | - | 3 | 1051 | |||
AK162194.1 | - | 3 | 1050 | |||
BC017161.1 | - | 3 | 1093 | 4481022 |