Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000035.3 | NP_000026.2 | 4 | 442 | ||
GenBank mRNA | AK026411.1 | - | 4 | 219 | ||
AK290795.1 | - | 4 | 436 | |||
BC029399.1 | - | 4 | 436 | 4593670 | ||
X01098.1 | - | 4 | 426 | |||
X02747.1 | - | 4 | 485 |