Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025948.2 | NP_080224.1 | 11 | 2062 | ||
GenBank mRNA | AK003611.1 | - | 9 | 1622 | ||
AK004427.1 | - | 1 | 195 | |||
AK012276.1 | - | 10 | 1825 | |||
AK013143.1 | - | 2 | 640 | |||
BC031521.1 | - | 11 | 2062 | 2655161 |