Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025959.3 | NP_080235.2 | 6, 7 | 473 | ||
GenBank mRNA | AK008996.1 | - | 217 | |||
AK012174.1 | - | 6, 7 | 474 | |||
AK014354.1 | - | 6, 7 | 461 | |||
AK144728.1 | - | 470 | ||||
AK166961.1 | - | 6, 7 | 458 | |||
BC025134.1 | - | 2, 3 | 161 | |||
BC037099.1 | - | 6, 7 | 471 | |||
BC043044.1 | - | 452 | ||||
BC057997.1 | - | 429 | ||||
BC116752.1 | - | 6, 7 | 473 | 40126071 | ||
BC119168.1 | - | 6, 7 | 473 | 8733917 |