Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025965.3 | NP_080241.3 | 8 | 1579 | ||
GenBank mRNA | AF326229.2 | - | 8 | 1554 | ||
AK010884.1 | - | 8 | 1526 | |||
AK013910.1 | - | 1 | 360 | |||
AK031538.1 | - | 8 | 1560 | |||
AK088181.1 | - | 1 | 189 | |||
AK160285.1 | - | 8 | 1542 | |||
AK166235.1 | - | 8 | 1548 | |||
AK167793.1 | - | 8 | 1521 | |||
BC011255.1 | - | 8 | 1510 | 4017600 |