Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025982.4 | NP_080258.1 | 6 | 790 | ||
GenBank mRNA | AK012457.1 | - | 6 | 791 | ||
AK012567.1 | - | 6 | 793 | |||
AK012853.1 | - | 6 | 793 | |||
AK014183.1 | - | 6 | 789 | |||
AK075962.1 | - | 6 | 791 | |||
AK160269.1 | - | 3 | 1479 | |||
AK211860.1 | - | 1 | 118 | |||
AK211955.1 | - | 1 | 118 | |||
AK214177.1 | - | 1 | 118 | |||
AK214294.1 | - | 1 | 118 | |||
AK215378.1 | - | 1 | 118 | |||
AK217758.1 | - | 1 | 119 | |||
AK218507.1 | - | 1 | 118 | |||
BC023306.1 | - | 6 | 767 | 4165774 |