Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000046.3 | NP_000037.2 | 2 | 1775 | ||
NM_198709.2 | NP_942002.1 | 3 | 844 | |||
XM_011543390.1 | XP_011541692.1 | 3 | 988 | |||
XM_011543391.2 | XP_011541693.1 | 2 | 551 | |||
XM_011543392.2 | XP_011541694.1 | 2 | 551 | |||
XM_011543393.1 | XP_011541695.1 | 2 | 551 | |||
XM_017009471.1 | XP_016864960.1 | 2 | 551 | |||
GenBank mRNA | AK290865.1 | - | 2 | 551 | ||
AK314903.1 | - | 2 | 560 | |||
BC029051.1 | - | 3 | 844 | 5186657 | ||
J05225.1 | - | 2 | 495 | |||
M32373.1 | - | 2 | 1048 |