Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000048.3 | NP_000039.2 | 14 | 1106 | ||
NM_001024943.1 | NP_001020114.1 | 13 | 1230 | |||
NM_001024944.1 | NP_001020115.1 | 12 | 1170 | |||
NM_001024946.1 | NP_001020117.1 | 12 | 1152 | |||
GenBank mRNA | BC008195.1 | - | 14 | 1068 | 3048146 | |
BC033146.1 | - | 13 | 1041 | 4101398 | ||
J03058.1 | - | 14 | 1064 | |||
KU177920.1 | - | 12 | 995 | |||
KU177921.1 | - | 11 | 917 | |||
KU177922.1 | - | 8 | 680 | |||
M14218.1 | - | 13 | 1109 | |||
M57638.1 | - | 13 | 1040 | |||
Y00753.1 | - | 13 | 1040 |