Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026034.4 | NP_080310.1 | 2, 3 | 409 | ||
GenBank mRNA | AK009821.1 | - | 2, 3 | 402 | ||
AK011303.1 | - | 2, 3 | 408 | |||
AK012872.1 | - | 2, 3 | 384 | |||
AK015272.1 | - | 2, 3 | 542 | |||
AK219320.1 | - | 1, 2 | 151 | |||
BC027343.1 | - | 2, 3 | 340 | |||
BC038487.1 | - | 2, 3 | 361 | 2812558 | ||
BC046281.1 | - | 2, 3 | 361 | 6395384 | ||
BC058573.1 | - | 2, 3 | 450 | 6487440 |