Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026058.4 | NP_080334.3 | 3 | 408 | ||
GenBank mRNA | AK013554.1 | - | 3 | 360 | ||
AK028937.1 | - | 3 | 334 | |||
AK028945.1 | - | 3 | 311 | |||
AK030280.1 | - | 3 | 348 | |||
AK036108.1 | - | 3 | 332 | |||
AK134887.1 | - | 3 | 365 | |||
AY029531.1 | - | 3 | 309 | |||
BC003946.1 | - | 3 | 295 | 3487343 | ||
CK617511.1 | - | 3 | 484 |