Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039045.1 | NP_001034134.1 | 2 | 627 | ||
NM_026078.2 | NP_080354.1 | 2 | 640 | |||
GenBank mRNA | AK008524.1 | - | 2 | 592 | ||
AK014096.1 | - | 2 | 602 | |||
AK016394.1 | - | 2 | 639 | |||
AK152660.1 | - | 2 | 618 | |||
AK155971.1 | - | 1 | 1549 | |||
AK161117.1 | - | 2 | 595 | |||
AK170153.1 | - | 2 | 595 | |||
BC006938.1 | - | 2 | 560 | 2649047 |