Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026083.2 | NP_080359.2 | 3 | 512 | ||
GenBank mRNA | AK013285.1 | - | 3 | 514 | ||
AK014200.1 | - | 3 | 292 | |||
AK078225.1 | - | 3 | 503 | |||
BB619434.1 | - | 3 | 513 | |||
BC138264.1 | - | 3 | 461 | 8861283 |