Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162523.1 | NP_001155995.1 | 2 | 348 | ||
NM_001162524.1 | NP_001155996.1 | 2 | 348 | |||
NM_026087.3 | NP_080363.2 | 2 | 348 | |||
GenBank mRNA | AK005400.1 | - | 2 | 350 | ||
AK005565.1 | - | 2 | 299 | |||
AK131656.1 | - | 2 | 338 | |||
AK145926.1 | - | 2 | 299 | |||
BC099443.1 | - | 2 | 286 | 30787251 |