Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007308.1 | - | 2 | 329 | ||
AK015125.1 | - | 2 | 305 | |||
BC043075.1 | - | 3 | 360 | 6404784 | ||
BC096688.1 | - | 2 | 278 |