Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025990.4 | NP_080266.2 | 9 | 1042 | ||
GenBank mRNA | AK009329.1 | - | 9 | 1038 | ||
AK009441.1 | - | 9 | 1044 | |||
AK009531.1 | - | 9 | 1038 | |||
AK009562.1 | - | 9 | 1038 | |||
AK009580.1 | - | 9 | 1042 | |||
AK009629.1 | - | 9 | 1041 | |||
AK009695.1 | - | 9 | 1039 | |||
AK009803.1 | - | 9 | 1040 | |||
AK009835.1 | - | 9 | 1042 | |||
AK010012.1 | - | 9 | 1042 | |||
AK010051.1 | - | 9 | 1038 | |||
AK010115.1 | - | 9 | 1037 | |||
AK010143.1 | - | 8 | 984 | |||
AK010145.1 | - | 9 | 1042 | |||
AK010158.1 | - | 8 | 959 | |||
AK010159.1 | - | 9 | 1041 |