Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025994.3 | NP_080270.2 | 1 | 333 | ||
GenBank mRNA | AK007560.1 | - | 1 | 323 | ||
AK011219.1 | - | 1 | 322 | |||
AK048475.1 | - | 1 | 334 | |||
AK150207.1 | - | 1 | 316 | |||
AK159367.1 | - | 1 | 323 | |||
AK167267.1 | - | 1 | 324 | |||
AK169409.1 | - | 1 | 323 | |||
AK195742.1 | - | 1 | 49 | |||
BC125303.1 | - | 1 | 365 | 40129818 |