Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026028.5 | NP_080304.1 | 3 | 341 | ||
GenBank mRNA | AK010251.1 | - | 3 | 194 | ||
AK012598.1 | - | 3 | 233 | |||
AK031590.1 | - | 3 | 429 | |||
AK166940.1 | - | 1 | 95 | |||
BC018387.1 | - | 3 | 169 | 3982552 | ||
CD555913.1 | - | 4 | 315 |