Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026040.3 | NP_080316.3 | 2, 3 | 191 | ||
GenBank mRNA | AK012865.1 | - | 2, 3 | 193 | ||
AK141882.1 | - | 185 | ||||
AK144845.1 | - | 190 | ||||
AK147692.1 | - | 169 | ||||
AY611629.1 | - | 2, 3 | 164 | |||
BC024338.1 | - | 168 | 5009612 |