Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam220a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026050.2 | NP_080326.2 | 3 | 1323 | ||
NM_133703.4 | NP_598464.3 | 2 | 1190 | |||
GenBank mRNA | AB374429.1 | - | 2 | 964 | ||
AK014805.1 | - | 3 | 1256 | |||
AK075875.1 | - | 2 | 1176 | |||
AK076127.1 | - | 2 | 1173 | |||
AK156614.1 | - | 3 | 1268 | |||
AY714985.1 | - | 1 | 739 | |||
BC005638.1 | - | 1 | 452 | |||
BC085297.1 | - | 2 | 1080 | |||
BC100440.1 | - | 2 | 1135 | |||
BC147186.1 | - | 2 | 792 | 9056217 | ||
BC147187.1 | - | 2 | 792 | 9056219 | ||
BC172003.1 | - | 2 | 792 | 9054289 |