Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026056.4 | NP_080332.1 | 7 | 875 | ||
GenBank mRNA | AK010235.1 | - | 7 | 622 | ||
AK013331.1 | - | 7 | 632 | |||
AK014979.1 | - | 3 | 271 | |||
AK033071.1 | - | 7 | 631 | |||
AK047701.1 | - | 5 | 442 | |||
AK076737.1 | - | 3 | 295 | |||
BC050752.1 | - | 7 | 636 | 6533812 | ||
BC057937.1 | - | 7 | 690 | 5325599 |