Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srek1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026075.2 | NP_080351.2 | 3 | 282 | ||
GenBank mRNA | AK013429.1 | - | 3 | 277 | ||
AK013737.1 | - | 3 | 296 | |||
AK014102.1 | - | 3 | 289 | |||
AK018357.1 | - | 3 | 307 | |||
AK020113.1 | - | 3 | 303 | |||
AK030777.1 | - | 2140 | ||||
AK135887.1 | - | 328 | ||||
BC027133.1 | - | 3 | 260 | 5359722 | ||
BC038302.1 | - | 3 | 315 | |||
BC096661.1 | - | 3 | 282 | 583227 | ||
BC116993.1 | - | 3 | 253 | 40126312 | ||
BC116999.1 | - | 3 | 253 | 40126318 |