Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026094.3 | NP_080370.2 | 3 | 232 | ||
GenBank mRNA | AK006684.1 | - | 3 | 228 | ||
AK018868.1 | - | 3 | 228 | |||
AK018877.1 | - | 3 | 228 | |||
AY364445.1 | - | 3 | 232 | |||
BC049570.1 | - | 3 | 198 | |||
BC118977.1 | - | 3 | 232 | 40098417 | ||
BC118978.1 | - | 3 | 232 | 40098419 |