Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026106.4 | NP_080382.2 | 2 | 1049 | ||
GenBank mRNA | AK007230.1 | - | 2 | 1024 | ||
AK018040.1 | - | 2 | 972 | |||
AK031057.1 | - | 2 | 983 | |||
AK075713.1 | - | 2 | 1049 | |||
AK090113.1 | - | 2 | 1031 | |||
AK139866.1 | - | 2 | 979 | |||
AK144749.1 | - | 2 | 1043 | |||
AK146573.1 | - | 2 | 1012 | |||
AK178588.1 | - | 2 | 269 | |||
AK211344.1 | - | 2 | 269 | |||
BC013461.1 | - | 2 | 618 | 4222118 |