Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025993.2 | NP_080269.1 | 3 | 2059 | ||
GenBank mRNA | AK010995.1 | - | 3 | 2060 | ||
AK077702.1 | - | 1 | 936 | |||
AK132010.1 | - | 3 | 2061 | |||
AK154531.1 | - | 3 | 2060 | |||
BC026790.1 | - | 3 | 1745 | 5148857 |