Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam103a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025997.2 | NP_080273.1 | 3 | 942 | ||
GenBank mRNA | AK011881.1 | - | 3 | 942 | ||
AK079819.1 | - | 3 | 921 | |||
AK081301.1 | - | 3 | 934 | |||
AK085230.1 | - | 3 | 940 | |||
AK150938.1 | - | 3 | 909 | |||
AK160347.1 | - | 3 | 941 | |||
AK169392.1 | - | 3 | 971 | |||
BC058367.1 | - | 3 | 865 | 6830048 | ||
BC091760.1 | - | 3 | 902 | 5042613 | ||
BC096399.1 | - | 3 | 900 | 876882 |
Gene Symbol: Gm10767 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177750.1 | NP_001171221.1 | 3 | 1241 | ||
GenBank mRNA | AK139034.1 | - | 3 | 1242 |