Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026021.3 | NP_080297.1 | 6 | 1174 | ||
GenBank mRNA | AJ409150.1 | - | 6 | 1135 | ||
AK005908.1 | - | 6 | 1162 | |||
AK012484.1 | - | 6 | 1175 | |||
AK088063.1 | - | 6 | 1195 | |||
BC027321.1 | - | 6 | 1140 | 3990966 |