Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2t | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278115.1 | NP_001265044.1 | 3 | 261 | ||
NM_026024.3 | NP_080300.1 | 3 | 438 | |||
GenBank mRNA | AK012565.1 | - | 3 | 397 | ||
AK021213.1 | - | 3 | 427 | |||
AK166098.1 | - | 3 | 408 | |||
BC029213.1 | - | 3 | 387 | 3591557 | ||
CK334733.1 | - | 3 | 252 |