Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026028.5 | NP_080304.1 | 10 | 1217 | ||
GenBank mRNA | AK012598.1 | - | 10 | 1109 | ||
AK031590.1 | - | 8 | 1021 | |||
BC018387.1 | - | 10 | 1045 | 3982552 |