Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026040.3 | NP_080316.3 | 3, 4 | 263 | ||
GenBank mRNA | AK012865.1 | - | 3, 4 | 265 | ||
AK141882.1 | - | 257 | ||||
AK144845.1 | - | 262 | ||||
AK147692.1 | - | 241 | ||||
AY611629.1 | - | 3, 4 | 236 | |||
BC024338.1 | - | 240 | 5009612 |