Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000089.3 | NP_000080.2 | 49 | 3872 | ||
GenBank mRNA | AK226074.1 | - | 49 | 3523 | ||
AL833478.1 | - | 6 | 4180 | |||
BC042586.1 | - | 49 | 3519 | 4803351 | ||
BC054498.1 | - | 49 | 3501 | 6474549 | ||
J03464.1 | - | 49 | 3870 | |||
KU177988.1 | - | 49 | 3401 | |||
V00503.1 | - | 18 | 1524 | |||
X55525.1 | - | 6 | 526 | |||
Z74616.1 | - | 49 | 3540 |