Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026056.4 | NP_080332.1 | 13 | 2285 | ||
GenBank mRNA | AK033071.1 | - | 13 | 2041 | ||
AK035556.1 | - | 13 | 2286 | |||
AK047701.1 | - | 11 | 1852 | |||
AK049813.1 | - | 10 | 1728 | |||
AK076737.1 | - | 9 | 1705 |